Search Thermo Fisher Scientific
Catalog Number | Width (English) Cabinet | Chamber Material | Sash Opening (English) | Electrical Requirements |
---|---|---|---|---|
1366-LED | 5 ft. | Stainless Steel (304) | 8 in. | 230 V, 50/60 Hz |
1366 | 5 ft. | Stainless Steel (304) | 8 in. | 230 V, 50/60 Hz |
Perform daily applications safely and efficiently with Thermo Scientific™ 1300 Series A2 Class II, Type A2 Biosafety Cabinets. These biological safety cabinets deliver excellent design and technology advancements such as superior protection with proprietary airflow design, exceptional ergonomics for a safe and comfortable environment and outstanding energy efficiency for operational cost savings.
Safety Features:
More Choices: