accuCap™ Capillary Junction pH Combination Electrodes - Glass Body, Gel-Filled Spear Tip - Mercury-Free
accuCap™ Capillary Junction pH Combination Electrodes - Glass Body, Gel-Filled Spear Tip - Mercury-Free
Actual product may vary
accuCap™ Capillary Junction pH Combination Electrodes - Glass Body, Gel-Filled Spear Tip - Mercury-Free
Large junction for increased flow, minimal clogging
Have Questions?
Catalog NumberConnector Types
13620133BNC
Catalog number 13620133
Price (SGD)
0.00
Each
Add to cart
Connector Types:
BNC
Price (SGD)
0.00
Each
Add to cart
Large junction for increased flow, minimal clogging
Better junction
  • Single-pore capillary junction is ∽200 times larger than typical ceramic junctions
  • Almost impossible to clog, even in difficult samples
  • Increased electrolyte flow, greater contact between sample and reference, fast and stable readings
Better reference design
  • Double junction design for enhanced performance
  • Diffusion barrier separates silver chloride reservoir from reference electrolyte
  • Permits use of silver-free electrolytes, prevents silver chloride loss from temperature-induced variations
  • Result: an extremely stable reference potential, providing long electrode life
Most accurate
  • Testing at German Federal Physical Technical Institute in Braunschweig determined this electrode design to be the most accurate laboratory electrode
  • See Traceability of pH measurement by Petra Spitzer; ISBN 3-89429-877-4 or SSN 0947-7063
  • Glass body and pH membrane
  • Rugged spear tip is ideal for soft foods
  • Polymer gel electrolyte provides reliable performance with difficult samples
Specifications
ColorPurple
Connector TypesBNC
JunctionDual Pore
MaterialGlass
pH Range2 to 14
Temperature (Metric) Operating0°C to 50°C
TypeCombination pH Electrode
Filling Solution TypePolymer gel
For Use With (Application)pH
For Use With (Equipment)pH meter
Temperature (English) Operating32°F to 122°F
Temperature (Metric) Range0°C to 50°C
Tip TypeSpear
Warranty1 Year
Length (English)1 in.
Length (Metric)25 mm
Diameter (English)0.23 in.
Diameter (Metric)6 mm
Unit SizeEach