accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
Actual product may vary
accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
Thermo Scientific
accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
For use on accumet™ Excel (XL) series benchtop meters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCell ConstantMeasurement Ranges
136201010.1 cm-10.5 to 200 μS/cm
13-620-1001 cm-110.0 to 2000 μS/cm
136201001 cm-110.0 to 2000 μS/cm
FIS13-620-1001 cm-110.0 to 2000 μS/cm
FSP13-620-1001 cm-110.0 to 2000 μS/cm
FSP136201001 cm-110.0 to 2000 μS/cm
FSP136201010.1 cm-10.5 to 200 μS/cm
13-620-10210 cm-11 to 200 mS/cm
FSP13-620-10210 cm-11 to 200 mS/cm
Catalog number 13620101
HSN 90279090
Price (SGD)
-
Cell Constant:
0.1 cm-1
Measurement Ranges:
0.5 to 200 μS/cm
For use on accumet™ Excel (XL) series benchtop meters
  • Immersion-type cell
  • Use in standard electrode holder: unique compact size is same as typical pH electrode
  • Built-in temperature sensors provide temperature-compensated readings
  • Epoxy body
Specifications
For Use With (Equipment)AB200, XL20, XL200, XL30, XL50, XL500, XL60, XL600 Benchtop Meters
Cell Constant0.1 cm-1
TypeConductivity Probe
Connector Types8-pin
MaterialEpoxy
Measurement Ranges0.5 to 200 μS/cm
Storage MethodImmersed in deionized water
Probe2 Cell Conductivity
Unit SizeEach