Nalgene™ Plastic Labware Value Pack
Nalgene™ Plastic Labware Value Pack
Actual product may vary
Nalgene™ Plastic Labware Value Pack
Nalgene™ Plastic Labware Value Pack
Nalgene™ Plastic Labware Value Pack
Nalgene™ Plastic Labware Value Pack
Enjoy an assortment of popular, everyday labware with the convenient Thermo Scientific™ Nalgene™ Labware Value Pack, perfect for a new lab starter or refresher pack.
Have Questions?
Catalog NumberCapacity (Metric)
1350-0001100 mL, 500 mL, 1 L (Graduated Cylinders), 50, 100, 250, 600, 1000 mL (Griffin Low Form Beakers), 500 mL (Straight-Sided Jars), 500 mL (Unitary Wash Bottles)
Catalog number 1350-0001
Price (SGD)
294.00
EA
Add to cart
Capacity (Metric):
100 mL, 500 mL, 1 L (Graduated Cylinders), 50, 100, 250, 600, 1000 mL (Griffin Low Form Beakers), 500 mL (Straight-Sided Jars), 500 mL (Unitary Wash Bottles)
Price (SGD)
294.00
EA
Add to cart
Choose from an assortment of popular, everyday labware with the Thermo Scientific™ Nalgene™ Plastic Labware Value Pack, perfect as a new lab starter or refresher pack. Nalgene Labware sets the standard for quality and superior performance for your valuable research with the highest quality materials for dependable performance.
Features:
  • Receive 20 items with one catalog number
  • A variety of everyday lab items at one low price
  • Won't shatter or break
Includes:
Six straight-sided jars (500mL/PPCO), four unitary wash bottles (500mL/LDPE), four graduated cylinders (2-100mL, 500mL, 1L/PPCO), five Griffin low form beakers (50, 100, 250, 600, 1000mL/PPCO), Thermo Scientific™ Nalgene™ Unwire™ blue half-rack for 30mm tubes.
Note:
Remove jar closure and autoclave separately. LDPE unitary wash bottles are not autoclavable. All other items in Value Pack are autoclavable.
Specifications
Capacity (Metric)100 mL, 500 mL, 1 L (Graduated Cylinders), 50, 100, 250, 600, 1000 mL (Griffin Low Form Beakers), 500 mL (Straight-Sided Jars), 500 mL (Unitary Wash Bottles)
MaterialPlastic
Item DescriptionLabware Value Pack
Certifications/ComplianceASTM E1272, CFR 21, ISO 6706, Part 177.1520
No. per Pack1
No. per Case1
Includes(4) Graduated Cylinders (2-100 mL, 500 mL, 1L/Ppco), (4) Unitary Wash Bottles (500 mL/Ldpe), (5) Griffin Low Form Beakers (50, 100, 250, 600, 1000 mL/Ppco), (6) Straight-Sided Jars (500 mL/Ppco), Unwire Blue Half-Rack for 30 mm Tubes
TypeLabware Value Pack
Unit SizeEA