Combination Wrench, 27mm
Combination Wrench, 27mm
Actual product may vary
Combination Wrench, 27mm
Ampco Safety Tools Combination Wrench, 27mm is non-sparking, non-magnetic and corrosion resistant
Have Questions?
Catalog NumberLength (Metric)
1338
also known as Q1338M
355.6 mm
Catalog number 1338
also known as Q1338M
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
355.6 mm
Made in USA
Specifications
For Use With (Equipment)Electronics, batteries, MRI, and precision magnetic equipment
Length (Metric)355.6 mm
MaterialAluminum Bronze
ColorBronze
For Use With (Application)Non-Sparking, Non-Magnetic, Corrosion Resistant Applications; Flammable or combustible vapors, liquids, dusts or residues; Corrosive environments like laboratories, acid, salt water and chemical processing
No. per Pack1 Ea.
Weight (Metric)410 g
Width (Metric)57.15 mm
Unit SizeEA