Combination Wrench
Combination Wrench
Actual product may vary
Combination Wrench
AMPCO™ Combination Wrench performs in high-risk and demanding environments.
Have Questions?
Catalog NumberSize
1336
also known as GT1336FS
26 mm
Catalog number 1336
also known as GT1336FS
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
26 mm
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
Specifications
MaterialAluminum Bronze
Size26 mm
Certifications/ComplianceFactory Mutual (FM) Approved
Unit SizeEA