accumet™ AB200 Benchtop pH/Conductivity Meters
Actual product may vary
accumet™ AB200 Benchtop pH/Conductivity Meters
accumet™ AB200 Benchtop pH/Conductivity Meters
accumet™ AB200 Benchtop pH/Conductivity Meters
accumet™ AB200 Benchtop pH/Conductivity Meters
Eutech
accumet™ AB200 Benchtop pH/Conductivity Meters
Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB200 pH/conductivity meter combines the benefits of a pH meter and conductivity meter in one easy to use, reliable measurement system. The large backlit display offers great visibility in laboratory environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
13-636-AB200pH/Conductivity Meter
FIS13-636-AB200pH/Conductivity Meter
FSP13-636-AB200pH/Conductivity Meter
Catalog number 13-636-AB200
Price (SGD)
-
Product Type:
pH/Conductivity Meter
Provides intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB200 pH/conductivity meter combines the benefits of a pH meter and conductivity meter in one easy to use, reliable measurement system. The large backlit display offers great visibility in laboratory environments.
  • Graphic backlit display with date and time, stability indicator and alarms
  • Dual channel meter for pH and conductivity measurements
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • Conductivity, TDS (total dissolved solids), salinity and resistivity modes
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (13-620-BSP) with five speed settings
NIST-Traceable Calibration Model (-CERT)
  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory, includes NIST-Traceable certificate
Specifications
  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Conductivity Range: 0.00μS to 500.0mS
  • Conductivity Resolution: 0.01, 0.1μS; 0.001, 0.01, 0.1mS
  • Conductivity Accuracy: ±1 % full scale
  • TDS Range: 0.00ppm to 500 ppt (TDS factor 1.00)
  • TDS Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • TDS Accuracy: ±1 % full scale
  • Salinity Range: 0 to 80.0 ppt
  • Salinity Resolution: 0.01, 0.1 ppm; 0.001, 0.01, 0.1 ppt
  • Salinity Accuracy: ±1 % full scale
  • Resistivity Range: 2.000 Ohm to 20.0 MOhm
  • Resistivity Resolution: 0.01, 0.1 Ohm; 0.001, 0.1 kOhm; 0.01 MOhm
  • Resistivity Accuracy: ±1 % full scale
  • Temperature Range: 0° to 100°C (32° to 212.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.3°C (±0.5°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, 8 Pin DIN, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial
Specifications
OutputsRS232 (phono plug), mini-B USB, stirrer
Accuracy (Conductivity)±1% FS
Accuracy (mV)±0.2 mV
Accuracy (pH)±0.002
Accuracy (Resistivity)± 1% full scale
Accuracy (Salinity)± 1% full scale
Accuracy (TDS)± 1% full scale
Calibration Points (Conductivity)Automatic (4 points), maximum 1/range, manual (5 points), maximum 1/range
Calibration Points (pH)Up to 6 preset or 5 custom
Cell Constant0.010 to 10.000
Certifications/ComplianceWithout NIST-traceable pH Certificate
Data Management DetailsManual, timed (selectable every 3 to 3600 sec.), printer or CSV format
Data Points500
Channels2
Display TypeLCD
Measurement ModespH, mV, Conductivity, TDS, Salinity, Resistivity, Temperature
Range (Conductivity)0 μS to 500 mS
Range (mV)±2000 mV
Range (pH)-2 to 20
Range (Resistivity)2 W to 20 MW
Range (Salinity)0 to 80 ppt
Range (TDS)0 ppm to 500 ppt (at TDS factor 1)
Resolution (pH)0.1/0.01/0.001
Temperature Range (Metric)0°C to 100°C
IncludesMeter, TRIS Compatible pH/ATC Electrode (13-620-631), Conductivity/Temp Probe (13-620-100), Electrode Arm, RS-232 and USB cables, 100/240V Power Supply
InputsBNC, ATC
ModelAB200 pH/Conductivity Kit
Product TypepH/Conductivity Meter
Resolution (Conductivity)0.01/0.1μS, 0.001/0.01/0.1mS
Resolution (mV)0.1mV
Resolution (Resistivity)0.01/0.1W, 0.001/0.1kW, 0.01MW
Resolution (Salinity)0.01/0.1ppm, 0.001/0.01/0.1ppt
Resolution (TDS)0/0.1ppm, 001/01/0.1ppt
Temperature Accuracy±0.3°C
Temperature Resolution0.1°C
Warranty3 Years
Voltage100/240 V
Frequency50/60Hz
Unit SizePer 100