accumet™ AB150 Benchtop pH Meters
Actual product may vary
accumet™ AB150 Benchtop pH Meters
accumet™ AB150 Benchtop pH Meters
accumet™ AB150 Benchtop pH Meters
accumet™ AB150 Benchtop pH Meters
accumet™ AB150 Benchtop pH Meters
Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
13-636-AB150BMeter with BioBasic Kit
FSP13-636-AB150BMeter with BioBasic Kit
FIS13-636-AB150Meter with pH Kit
FSP13-636-AB150Meter with pH Kit
FIS13-636-AB150ApH Meter
FKS12840633pH Meter
FSP13-636-AB150ApH Meter
Catalog number 13-636-AB150B
Price (SGD)
-
Type:
Meter with BioBasic Kit
Provide intuitive, simple operation and high accuracy measurements in a compact, affordable meter. Fisherbrand accumet AB150 pH benchtop meters have large backlit display which offers great visibility in laboratory environments. The accumet AB150 pH meter is an industry favorite for when reliability and consistency in measurements matter.
  • Graphic backlit display with date and time, stability indicator and alarms
  • One to six point pH calibration with 0.001 pH resolution
  • Automatic buffer recognition with four pH buffer sets or custom buffer option
  • pH slope and offset display
  • One point relative mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation, displayed in °C or °F
  • Password protection of calibration and setup modes
  • 500 data set storage
  • RS232 and USB interfaces
  • Multilanguage interface with English, German, Chinese, French, Italian and Spanish
  • Optional stirring probe (13-620-BSP) with five speed settings
NIST-Traceable Calibration Models (-CERT)
  • Help meet ISO, FDA, USDA and EPA quality assurance guidelines
  • Includes meter accuracy at five test points across pH range
  • Includes meter accuracy at five test points across corresponding mV range
  • Testing performed by accredited laboratory; includes NIST-Traceable certificate
Specifications
  • pH Range: -2.000 to 20.000 pH
  • pH Resolution: 0.1, 0.01, 0.001 pH
  • pH Accuracy: ±0.002 pH
  • mV Range: ±2000.0mV
  • mV Resolution: 0.1mV
  • mV Accuracy: ±0.2mV or ±0.05 %, whichever is greater
  • mV Offset Adjustment: Up to ±150mV
  • Temperature Range: 0° to 100°C (32° to 212.0°F)
  • Temperature Resolution: 0.1°C (0.1°F)
  • Temperature Accuracy: ±0.3°C (±0.5°F)
  • Calibration Points: 1 to 6 points with automatic buffer recognition (ABR)
  • ATC Temperature Calibration: Offset in 0.1°C increments; Offset range: ±5°C (±9°F)
  • Self-Test: Yes
  • Inputs: BNC, mini-phone ATC, ground
  • Power Requirements: 100 to 240VAC, 50/60Hz
Applications
Academic, research, QA/QC, drinking water, wastewater, food, beverage, industrial
Includes:
13-636-AB150A: accumet AB150 pH meter, electrode stand, RS232 and USB cables, 100/240V universal power supply.
13-636-AB150: accumet AB150 pH meter, TRIS-compatible double junction refillable epoxy-body pH/ATC electrode (13-620-631), electrode stand, RS232 and USB cables, 100/240V universal power supply.
13-636-AB150B: accumet AB150 pH meter, TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC temp probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V universal power supply.
AB150ACERT: accumet AB150 pH meter, electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST certificate.
AB150CERT: accumet AB150 pH meter, TRIS-compatible double junction refillable epoxy-body pH/ATC Electrode (13-620-631), electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST certificate.
AB150BCERT: accumet AB150 pH meter, TRIS-compatible double junction refillable glass-body pH electrode (13-620-183A), ATC temp probe (13-620-19), electrode stand, RS232 and USB cables, 100/240V universal power supply, NIST certificate.
Specifications
Accuracy (Temperature)±0.3°C
Accuracy (mV)±0.2
Accuracy (pH)±0.002
Data Management DetailsManual, Timed (selectable every 3 to 3600 sec.), Printer or CSV Format
OutputsRS-232 (phono plug), mini-B USB, stirrer
IncludesMeter, TRIS Compatible accuTupH pH Electrode (13-620-183A), ATC Probe (13-620-19), Electrode Arm, 110/220V Power Supply
Range (pH)-2 to 20
Resolution (pH)0.001, 0.01, 0.1
Calibration Points (pH)Up to 6 pre-set or 5 custom
Range (mV)±2000
Resolution (mV)0.1 mV
Calibration Points1 to 6
Temperature Range (Metric)0°C to 100°C
Resolution (Temperature)0.1°C
Display TypeLCD
Data Points500
InputsBNC, ATC
Certifications/ComplianceWithout NIST-traceable pH Certificate
Warranty3 Years
ModelAB150 pH BioBasic Kit
TypeMeter with BioBasic Kit
Voltage100/240 V
Frequency50/60 Hz
Unit SizePer 100