accumet™ Epoxy Body Mercury-Free Reference Electrode
Actual product may vary
accumet™ Epoxy Body Mercury-Free Reference Electrode
A rugged epoxy body makes this a great choice for demanding environments
Have Questions?
Catalog NumberConnector Types
13-620-858Pin
Catalog number 13-620-858
Price (SGD)
-
Connector Types:
Pin
A rugged epoxy body makes this a great choice for demanding environments
  • Rugged epoxy body for in-plant monitoring, field measurements, instructional labs, classrooms
  • Stable with ion-selective electrodes
  • Tough enough to hand hold, use directly in a vessel, stir solutions
  • No need for auxiliary stirrer
Use with indicating half cell.
Specifications
Connector TypesPin
JunctionDouble, Porous Polymer
MaterialEpoxy
Temperature (Metric) OperatingMax 80°C
TypeMercury-Free Reference Electrode
Filling Solution TypeLiquid
For Use With (Application)Field, lab
Length (English) Cable30 in.
Length (Metric)106 mm
Diameter (Metric)12 mm
RefillableRefillable
Unit SizeEach