Check Order Status
VaporTrap™ Polypropylene, 83mm Carboy Caps with Filter Kit, Four Ports 1/8 in. OD Tubing, Three Ports 1/4 in. OD Tubing
Actual product may vary
VaporTrap™ Polypropylene, 83mm Carboy Caps with Filter Kit, Four Ports 1/8 in. OD Tubing, Three Ports 1/4 in. OD Tubing
For Use With:
carboys with 83mm cap
Have Questions?
Catalog Number
Description
12826
also known as WM1282X6
Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports
Catalog number
12826
also known as WM1282X6
Price (SGD)
303.00
EA
Add to cart
Description:
Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports
Price (SGD)
303.00
EA
Add to cart
Carbon filter prevents volatile organic compound vapors
Specifications
Description
Four (4) 1/8 in. Tubing OD Ports and Three (3) 1/4 in. Tubing OD Ports
Dimensions (L x W x H)
5 x 5.5 x 11 in.
For Use With (Equipment)
carboys with 83mm cap
Material
Polypropylene
Product Type
Cap