Samco™ Clicktainer™ Vials and Specimen Containers
Samco™ Clicktainer™ Vials and Specimen Containers
Actual product may vary
Samco™ Clicktainer™ Vials and Specimen Containers
Thermo Scientific™ Samco™ Clicktainer Vials and Specimen Containers provide excellent sample protection and user safety by ensuring correct cap application the every time.
Have Questions?
Catalog NumberCapacity (Metric)Closure ColorProduct TypeLabel
120WHT53-1000120 mLWhitePre-assembled Clicktainer VialTabbed
Catalog number 120WHT53-1000
Price (SGD)
272.00
CS
Add to cart
Capacity (Metric):
120 mL
Closure Color:
White
Product Type:
Pre-assembled Clicktainer Vial
Label:
Tabbed
Price (SGD)
272.00
CS
Add to cart
Providing sample protection and user safety with the Thermo Scientific™ Samco™ Clicktainer™ Vials and Specimen container, equipped with an audible multi-click seal. Help enable improved patient diagnosis with a leak tight seal, easy to grip surfaces and straight sided design for reliable and safe sample management.
Superior Leakproof Performance
  • Proven leakproof in accordance with BS EN 14254
  • Validated as 95kPa compliant at ambient temperature
  • Rigorously tested for suitability in pneumatic tubes
Clearly Designed Labels
  • One label multiple languages –English, Spanish and French
  • Fields to capture all FDA required information
  • Optional temperature strip
  • Also available unlabeled
Wide Range of Cap Colors and Packaging Configurations
  • Cap colors to differentiate and manage samples
  • Available pre-assembled, sterile and non-sterile, for convenient usage
  • Separate closure and vial bulk packs for fixative pre-filling applications
  • Individual peel pouches for operating rooms, cleanrooms, and other sterile environments
  • Dispenser boxes ideal for neat storage in smaller labs and doctors’ offices
† The term leakproof applies to Thermo Scientific Samco Clicktainer products, 90mL/53mm and 120mL/53mm, that meet the following criteria: a) a vacuum test that draws 95 kPa of pressure within a sealed chamber with no leakage; b) leak testing in accordance with BS EN 14254, which involves filling the container with a dye, placing for 2 minutes on a roller mixer followed by 1 hour inverted in a water bath with no leakage. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Samco Clicktainer Vials under conditions of their planned applications. Thermo Scientific Samco Clicktainer Vials are leakproof at ambient temperature and pressure when used with their Clicktainer closures.
Specifications
Product TypePre-assembled Clicktainer Vial
Closure IncludedYes
MaterialPolypropylene
SterilitySterile
Capacity (English)4 oz.
Capacity (Metric)120 mL
Certifications/Compliance95 kPA
Closure ColorWhite
Closure MaterialHDPE
ColorClear
DescriptionThermo Scientific Samco Clicktainer Vial, 120 mL, 53 mm diameter, Tabbed Label, White Cap, Sterile, Assembled
Diameter (Metric)53 mm
IncludesPre-assembled Clicktainer Vials with White Caps (120 mL)
LabelTabbed
No. per Case300/Cs.
No. per Pack75/Pk.
Unit SizeCS