Fiberlite F23-48 x 1.5 for LYNX 4000 and 6000 Superspeed Centrifuges
Fiberlite F23-48 x 1.5 for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
Fiberlite F23-48 x 1.5 for LYNX 4000 and 6000 Superspeed Centrifuges
Combine high g-force and high capacity for 1.5mL microtubes with the Thermo Scientific™ Fiberlite™ F23-48 x 1.5mL Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
096-48407548 x 1.5 mL
Catalog number 096-484075
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
48 x 1.5 mL
Combine high g-force and high capacity for 1.5mL microtubes with the Thermo Scientific™ Fiberlite™ F23-48 x 1.5mL Fixed-Angle Rotor.
Features:
  • Lightweight carbon fiber and innovative construction allow unique rotor designs not possible with metal
  • Corrosion and fatigue-resistance secures the rotor's structural integrity for unequalled durability
  • Rotor is backed by a 15-year warranty
Thermo Scientific™ Auto-Lock™ Rotor Exchange:
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification:
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle:
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionFiberlite F23-48x1.5 Fixed Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity48 x 1.5 mL
Max. RCF57,368 x g
Max. Speed23,000 rpm
Radius (Metric)97 mm
Radius (Metric) Min.64 mm
K Factor137
Max. Tube Length40 mm
Max. Tube Diameter11 mm
Net Weight10.0 lb. (4.54 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle45°
Unit SizeEA