Fiberlite F37L-8 x 100 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Fiberlite F37L-8 x 100 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Actual product may vary
Fiberlite F37L-8 x 100 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Reduce the required number of runs for high-volume separations with this ultraspeed carbon fiber rotor that features two additional tube cavities.
Have Questions?
Catalog NumberCapacity
096-0870568 x 100 mL
Catalog number 096-087056
Price (SGD)
45,069.00
EA
Add to cart
Capacity:
8 x 100 mL
Price (SGD)
45,069.00
EA
Add to cart
Reduce the required number of runs for high-volume separations with the Thermo Scientific™ Fiberlite™ F37L-8 x 100 Fixed-Angle Rotor, an ultraspeed carbon fiber rotor that offers two additional tube cavities for an eight-place total capacity.
Fiberlite rotors:
  • Reduce run-times with higher G-forces achieved with lightweight carbon fiber rotors
  • Ensure structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Ordering information:
Tube adapters are sold separately accommodating a variety of tubes in different capacities. Adapters available in packages of two. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ WX Ultracentrifuge Series and Beckman™ Floor Model L Series (except Optima) centrifuges, Nalgene™ bottles and tubes
Recommended for:
Separations of subcellular organelles or concentration of viruses.
Specifications
DescriptionFiberlite F37L-8 x 100 Fixed-Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity8 x 100 mL
Max. RCF182,460 x g
Max. Speed37,000 rpm
Radius (Metric)119 mm
Radius (Metric) Min.48.8 mm
K Factor168
Max. Tube Length102 mm
Max. Tube Diameter38 mm
Net Weight25 lb. (11.34 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle25°
Min. RCF73,600 x g
Unit SizeEA