Fiberlite F65L-6 x 13.5 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Fiberlite F65L-6 x 13.5 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Actual product may vary
Fiberlite F65L-6 x 13.5 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Process 6 x 13.5mL at speeds of 65,000rpm and maximum RCF of 324,000 x g with the corrosion-free carbon fiber Thermo Scientific™ Fiberlite™ F65L-6 x 13.5 Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
096-0671356 x 13.5 mL
Catalog number 096-067135
Price (SGD)
29,565.00
EA
Add to cart
Capacity:
6 x 13.5 mL
Price (SGD)
29,565.00
EA
Add to cart
Process 6 x 13.5mL at speeds up to 65,000rpm and maximum RCF of 324,000 x g with the corrosion-free carbon fiber Thermo Scientific™ Fiberlite™ F65L-6 x 13.5 Fixed-Angle Rotor.
Fiberlite rotors:
  • Reduce run-times with higher G-forces with our lightweight carbon fiber rotors
  • Ensure structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Ordering information:
Tube adapters are sold separately to accommodate a variety of tubes varying in different capacities; available in packages of two. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ WX, Sorvall Discovery™ SE Series and Beckman™ Optima L Series centrifuges, Nalgene™ bottles and tubes
Recommended for:
Small capacity density gradient centrifugation of sub cellular particles, nucleic acids and differential centrifugation of small particles
Specifications
DescriptionFiberlite F65L-6 x 13.5 Fixed-Angle Rotor
MaterialCarbon Fiber Composite
TypeFixed Angle Rotor
Capacity6 x 13.5 mL
Max. RCF324,000 x g
Max. Speed65,000 rpm
Radius (Metric)68.4 mm
Radius (Metric) Min.21 mm
K Factor70
Max. Tube Length76 mm
Max. Tube Diameter16 mm
Net Weight9.5 lb. (4.31 kg)
For Use With (Equipment)Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle30°
Min. RCF99,372 x g
Unit SizeEA