Point of Dispense Filters for Barnstead™ Point of Use Water Purification Systems
Actual product may vary
Point of Dispense Filters for Barnstead™ Point of Use Water Purification Systems
Sterile filter for use with Barnstead™ water purification systems; can be autoclaved up to 5 times.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
09.1003Sterile Filter, 0.2μm
50157375Sterile Filter, 0.1μm
FIS10-451-026Sterile Filter, 0.2μm
FKS11367111Sterile Filter, 0.2μm
Catalog number 09.1003
HSN 84212190
Price (SGD)
368.00
Each
Description:
Sterile Filter, 0.2μm
Price (SGD)
368.00
Each
Reduce bacteria using the Thermo Scientific™ Barnstead™ Point of Use Water Purification Systems.
Replacement consumable for the following Barnstead water purification systems:
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • LabTower™ EDI
  • LabTower™ TII
  • MicroPure™
  • Smart2Pure™
  • Smart2Pure™ Pro
Specifications
DescriptionSterile Filter, 0.2μm
For Use With (Equipment)GenPure, GenPure xCAD, LabTower EDI, GenPure Pro
TypePoint of Dispense Filter
Unit SizeEach