Biomechanical Kits
Biomechanical Kits
Actual product may vary
Biomechanical Kits
Biomechanical Kits
Biomechanical Kits
Biomechanical Kits
Biomechanical Kits
Offer a complete lab-tested system for teaching and learning the musculoskeletal system
Have Questions?
Catalog NumberProduct TypeIncludes
06.5015Complete Biomechanics Module KitBiomechanical arm and leg models, support base, rod and clamps, force cord assembley (cable, handle connectors, fine adjustment force gauge), brass weights (1 -50g, 1-100 g, 2-200g), instructor's curriculum guide and four student laboratory guides
Catalog number 06.5015
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Complete Biomechanics Module Kit
Includes:
Biomechanical arm and leg models, support base, rod and clamps, force cord assembley (cable, handle connectors, fine adjustment force gauge), brass weights (1 -50g, 1-100 g, 2-200g), instructor's curriculum guide and four student laboratory guides
By mastering a limited set of basic concepts, students can predict and explain body movement using the same approaches employed by kinesiologists, exercise physiologists, physical therapists, and orthopedic nurses. Working ideally in teams of four, students practice observation, measurement, data gathering and analysis, prediction, plus calculations and problem solving.
Specifications
Product TypeComplete Biomechanics Module Kit
For Use With (Application)For teaching and learning the musculoskeletal system
IncludesBiomechanical arm and leg models, support base, rod and clamps, force cord assembley (cable, handle connectors, fine adjustment force gauge), brass weights (1 -50g, 1-100 g, 2-200g), instructor's curriculum guide and four student laboratory guides
MountingSupport Base and Rod
Unit SizeEA