▸
Water Purification
▸
Water Purification Accessories and Replacement Parts
▸
CO
2
Adsorber + Sterile Filter, 0.2μm
Actual product may vary
CO
2
Adsorber + Sterile Filter, 0.2μm
Save water system cartridge life by preventing CO
2
from entering the water tank with the combination Thermo Scientific™ CO
2
Adsorber + Sterile Filter.
Have Questions?
Catalog Number
For Use With (Equipment)
06.5002
LabTower EDI, Pacific TII, Smart2Pure, Pacific RO
Catalog number
06.5002
HSN
84213920
Price (SGD)
571.00
Each
Add to cart
For Use With (Equipment):
LabTower EDI, Pacific TII, Smart2Pure, Pacific RO
Price (SGD)
571.00
Each
Add to cart
Save water system cartridge life by preventing CO
2
from entering the water tank with the combination Thermo Scientific™ CO
2
Adsorber + Sterile Filter.
Use with the Following Thermo Scientific™ Barnstead™ Water Systems:
LabTower™ EDI
Pacific™ RO
Pacific™ TII
Smart2Pure™ (12 System only)
Specifications
Description
CO2 adsorber + sterile filter, 0.2µm
For Use With (Equipment)
LabTower EDI, Pacific TII, Smart2Pure, Pacific RO