1x1 image pixel for data collection
Buffer Solution pH 10.01
Buffer Solution pH 10.01
Actual product may vary
Buffer Solution pH 10.01
Buffers and calibration solutions packed in durable plastic bottles – economical and convenient for laboratory use. ECBU10BTC
Have Questions?
Catalog number 01X211205
Price (SGD)
-
Buffers and calibration solutions packed in durable plastic bottles – economical and convenient for laboratory use. ECBU10BTC
  • Buffers and calibration solutions
  • Packed in 480 mL durable plastic bottles
  • Economical and convenient for laboratory use
Specifications
CAS497-19-8
Boiling PointApproximately 100°C
ColorBlue
CAS3844-45-9
CAS144-55-8
For Use With (Application)Measuring pH
CAS99-76-3
For Use With (Equipment)pH Meter
pH10
Physical FormLiquid
Solution TypeAqueous
CAS7732-18-5
FormulationSodium carbonate, Deionized water, Methyl Paraben, Erioglaucine
PackagingPlastic Bottle
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link