Check Order Status
Buffer Solution pH 4.01
Buffer Solution pH 4.01
Buffer solution pH 4.01 red 480mL ECBU4BTC
Have Questions?
Catalog Number
Quantity
01X211204
480 mL
Catalog number
01X211204
Price (SGD)
-
Quantity:
480 mL
Buffer solution pH 4.01 red 480mL ECBU4BTC
Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
Color
Red
pH
4.01
Quantity
480 mL