Fiberlite 500mL Bottle
Fiberlite 500mL Bottle
Actual product may vary
Fiberlite 500mL Bottle
Thermo Scientific™ 500mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottles are available for Thermo Scientific™ Sorvall™ LYNX Superspeed
Have Questions?
Catalog NumberMaterial
010-1493Polypropylene
Catalog number 010-1493
Price (SGD)
494.00
EA
Add to cart
Material:
Polypropylene
Price (SGD)
494.00
EA
Add to cart
Thermo Scientific™ 500mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottles will be available for Thermo Scientific™ Sorvall™ LYNX Superspeed
Features
  • PPCO or PC
  • Increased cycle life compared to previous Thermo Scientific bottles - 100 cycle life (caps + bottles)
  • Certifications USP Class VI - Non-Cytotoxic
  • Max speeds match rotors
  • Ergonomic speed handle design allows much easier bottle handling and removal
Specifications
Capacity (Metric)500 mL
Capacity (English)16.9 oz.
ColorClear
IncludesGas chromatograph, split/splitless inlet, FID, auxiliary valve oven, 4-port valve, two 6-port valves, and columns
MaterialPolypropylene
Product LineFiberlite
Volume (Metric) Brim500 mL
Certifications/ComplianceUSP Class VI Non-Cytotoxic
GraduatedNo
ShapeRound
Product TypeCentrifuge Bottle
Closure MaterialPolypropylene
Closure TypeSealing Cap
Through the Closure DispensingYes
Unit SizeEA
Frequently asked questions (FAQs)
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.