Fiberlite™ F14-6 x 250y Adapters
Actual product may vary
Fiberlite™ F14-6 x 250y Adapters
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support various conical tubes.
Have Questions?
Catalog NumberCapacity
010-141015 mL Conical Tube
Catalog number 010-1410
HSN 39269099
Price (SGD)
959.00
Special offer
Ends: 30-Jun-2026
1,128.00
Save 169.00 (15%)
PK
Capacity:
15 mL Conical Tube
Price (SGD)
959.00
Special offer
Ends: 30-Jun-2026
1,128.00
Save 169.00 (15%)
PK
Add versatility to your fixed-angle rotor with Thermo Scientific™ Fiberlite™ F14-6 x 250y Adapters, which support Ridge™, Vacutainer™ and conical tubes.
Adapters hold Oak Ridge™, Vacutainer™ and conical tubes, and vary by capacity.
Ordering Information:
Order Lid O-ring (022062034), O-ring refresher kit (021062038), Lid assembly (099062034) and Fiberlite F14-6x250y Rotor (78500) separately. Contact your Sales Representative for more information.
Specifications
TypeRotor Tube Adapter
Capacity15 mL Conical Tube
Vessels per Adapter5
Unit SizePK