Check Order Status
Ferrous Sulfate Heptahydrate, >99%, (Crystalline), MP Biomedicals
Element Search
Structure Search
MP Biomedicals
Ferrous Sulfate Heptahydrate, >99%, (Crystalline), MP Biomedicals
Have Questions?
Catalog Number
Quantity
MPB0215804883
250 g
Catalog number
MPB0215804883
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material
Ferrous Sulfate Heptahydrate
CAS
7782-63-0
Recommended Storage
Room Temperature
Physical Form
Crystalline
Quantity
250 g
View more