Copper(II) sulfate pentahydrate, 99%
Copper(II) sulfate pentahydrate, 99%
Copper(II) sulfate pentahydrate, 99%
Copper(II) sulfate pentahydrate, 99%
Copper(II) sulfate pentahydrate, 99%
Thermo Scientific Chemicals
Copper(II) sulfate pentahydrate, 99%
CAS: 7758-99-8 | CuH10O9S | 249.68 g/mol
Have Questions?
Catalog NumberQuantity
FSKA11262.0B1000 g
Catalog number FSKA11262.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper (II) sulfate pentahydrate
CAS7758-99-8
Health Hazard 1H302-H315-H319
Health Hazard 2GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
pH4
View more
Copper(II) sulfate, pentahydrate can be added with potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Copper(II) sulfate, pentahydrate can be added with potassium permanganate to give an oxidant for the conversion of primary alcohols. It is used to etch zinc or copper plates for intaglio printmaking and as a mordant in vegetable dyeing. In analytical chemistry, it is used in Fehling′s solution and Benedict′s solution to test for reducing sugars. Also, used to test protein in Biuret test.
Solubility
Soluble in water and methanol. Insoluble in alcohol.
Notes
Incompatible with steel, finely powdered metals, hydroxylamine, magnesium, hydrazine, nitromethane, alkalies, phosphates and acetylene gas
RUO – Research Use Only
General References:
  1. Ford, D. D.; Lenahan, S.; Jorgensen, M.; Dube, P.; Delude, M.; Concannon, P. E.; Anderson, S. R.; Oyler, K. D.; Cheng, G.; Mehta, N.; Salan, J. S. Development of a Lean Process to the Lead-Free Primary Explosive DBX-1. Org. Process Res. Dev. 2015, 19 (6), 673-680.
  2. Golf, H. R. A.; Reissig, H.; Wiehe, A. Synthesis of SF5-Substituted Tetrapyrroles, Metalloporphyrins, BODIPYs, and Their Dipyrrane Precursors. J. Org. Chem. 2015, 80 (10), 5133-5143.