Pipettor Tip Brushes
Pipettor Tip Brushes
Actual product may vary
Fisher Catalogue
Pipettor Tip Brushes
Features tapered bristles. Pinnacle Brush™ Pipettor Tip Brushes are for use with pipets.
Have Questions?
Catalog NumberSize
FKS12801954Tapered: 13 to 3 mm
Catalog number FKS12801954
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Size:
Tapered: 13 to 3 mm
  • Metal handle
  • Nylon bristles
Specifications
Product TypePipettor Tip Brush
Bristle MaterialNylon
For Use With (Equipment)Pipettor
Length (Metric)230 mm
Quantity5 Pk.
SizeTapered: 13 to 3 mm
Unit SizePK