PET Storage Bottles, Octagonal, Sterile
PET Storage Bottles, Octagonal, Sterile
Actual product may vary
Fisher Catalogue
PET Storage Bottles, Octagonal, Sterile
Manufactured with polyethylene terephthalate (PET) for a lightweight, break resistant and crystal clear product. Corning™ PET Storage Bottles, Octagonal are ideal for storage and sampling of media, biologicals and other aqueous solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS11761595250 mL50 to 250 mL
FIS07-201-425125 mL25 to 125 mL
FIS07-201-426250 mL50 to 250 mL
FIS07-201-427500 mL50 to 500 mL
FKS11721595500 mL50 to 500 mL
FKS117415951 L50 to 1000 mL
Catalog number FKS11761595
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
250 mL
Graduations:
50 to 250 mL
Polyethylene terephthalate (PET) bottles are ideal for storage and sampling of media, biologicals, and other aqueous solutions. The PET resin makes the bottle lightweight, break resistant and as transparent as glass.
Features:
  • Packaged in two convenient, shrink-wrapped trays within an outer bag to assure cleanliness
  • Break-resistant bottles are an ergonomic and easy to handle alternative to glass
  • Bottles are sterilized to 10−6 SAL and certified nonpyrogenic.
  • Leakproof, tamper-evident 18mm HDPE screw cap
  • Molded graduations for accurate measurements
  • USP class VI compliant bottles are validated noncytoxic and nonhemolytic
Specifications
Capacity8.45 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorNatural
Diameter (English) Inner Neck1.2 in.
Diameter (Metric) Inner Neck31.7 mm
For Use With (Application)For storage and sampling of media, biologicals and other aqueous solution
MaterialPET
No. per Case48/Cs.
No. per Pack24/Pk.
Quantity24 Pk.
SterilitySterile
Closure ColorNatural
Graduations50 to 250 mL
Neck Finish34.7 mm
Certifications/ComplianceUSP Class VI
GraduatedYes
MouthWide
ShapeOctagonal
Height (Metric)142 mm
Height (English)5.5 in.
Closure MaterialHDPE
Closure Size31.7 mm
Closure TypeScrew Cap
Unit SizePK