Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Length (Metric) | Diameter (Metric) |
---|---|---|---|
FKS11597403 | 36 mL | 150 mm | 20 mm |
FIS14-961-25 | 4 mL | 75 mm | 10 mm |
FKS11517403 | 6 mL | 75 mm | 12 mm |
FIS14-961-26 | 6 mL | 75 mm | 12 mm |
FIS14-961-27 | 10 mL | 100 mm | 13 mm |
FKI14-961-27 | 10 mL | 100 mm | 13 mm |
FKS11527403 | 10 mL | 100 mm | 13 mm |
FIS14-961-28 | 13 mL | 85 mm | 15 mm |
FIS14-961-29 | 15 mL | 100 mm | 16 mm |
FIS14-961-30 | 19 mL | 125 mm | 16 mm |
FIS14-961-31 | 23 mL | 150 mm | 16 mm |
FIS14-961-32 | 28 mL | 150 mm | 18 mm |
FKS11557403 | 19 mL | 125 mm | 16 mm |
FIS14-961-33 | 36 mL | 150 mm | 20 mm |
FIS14-961-34 | 55 mL | 150 mm | 25 mm |
FKS11507413 | 55 mL | 150 mm | 25 mm |