General-Purpose Narrow-Neck Bottles
General-Purpose Narrow-Neck Bottles
Actual product may vary
Fisher Catalogue
General-Purpose Narrow-Neck Bottles
Use these general purpose bottles for liquid storage and sampling. Fisherbrand™ General-Purpose Narrow-Neck Bottles offer excellent chemical resistance. Bottles can be recycled.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS115874331000 mL
FKS11557433125 mL
FKS11567433250 mL
FKS11577433500 mL
Catalog number FKS11587433
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
  • Clear general purpose bottles
  • Polypropylene screw closure
Specifications
Capacity (Metric)1000 mL
ColorTranslucent
For Use With (Application)For liquid storage and sampling
MaterialPolypropylene
Closure IncludedYes
MouthNarrow
Product TypeWash Bottle
Closure MaterialPolypropylene
Closure Size28/R6
Diameter (Metric)83 mm
Closure TypeScrew Cap
Unit SizePK