Check Order Status
Reusable Polyethylene Dippers
Actual product may vary
Reusable Polyethylene Dippers
500mL beakers with long handles for removing samples from large tanks.
Have Questions?
Change view
Catalog Number
Length (Metric) Handle
Length (English) Handle
FIS14-242-1
0.9 m
3 ft.
FIS14-242-10
3.7 m
12 ft.
FIS14-242-5
1.8 m
6 ft.
3 Options
Catalog number
FIS14-242-1
Price (SGD)
132.00
Each
Add to cart
Length (Metric) Handle:
0.9 m
Length (English) Handle:
3 ft.
Price (SGD)
132.00
Each
Add to cart
500mL beakers with long handles for removing samples from large tanks.
Lightweight, durable polyethylene
Graduated beaker features dual pouring spouts for easy left- or right-handed operation
Handle has an “easy-grip” end for secure handling under adverse conditions
Specifications
Product Type
Dipper
Material
HDPE
Capacity (Metric)
500 mL
For Use With (Application)
For removing samples from large tanks
Dimensions (L x W x H)
5.75 x 5 x 43 in. (14.6 x 12.7 x 109.22 cm)
Height (English)
43 in.
Height (Metric)
109.22 cm
Length (English)
5.75 in.
Length (English) Handle
3 ft.
Length (Metric)
14.6 cm
Length (Metric) Handle
0.9 m
Width (English)
5 in.
Width (Metric)
12.7 cm