Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Actual product may vary
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Standard Class B Graduated Cylinders
Calibrated To Contain
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsLimit of Error
FIS08-557-1F500 mL5 mL+/- 4.0mL
FIS08-557-1A10 mL0.2 mL+/- 0.20mL
FIS08-557-1D100 mL1 mL+/- 1.0mL
FIS08-557-1E250 mL2 mL+/- 2.0mL
FIS08-557-1G1000 mL10 mL+/- 6.0mL
Catalog number FIS08-557-1F
Price (SGD)
132.00
EA
Add to cart
Capacity (Metric):
500 mL
Graduations:
5 mL
Limit of Error:
+/- 4.0mL
Price (SGD)
132.00
EA
Add to cart
Calibrated To Contain
  • White-ceramic graduations
  • Heavy round bases (except 2000mL size, which has molded hexagonal base)
  • Handy pouring spouts
  • Accuracy is assured even at the lowest graduations
Specifications
Base TypeHexagonal
CalibrationTo Contain
Scale ColorWhite
ShapeRound
Certifications/ComplianceASTM D447, ASTM D86, ASTM E133
ClassB
Height (English)14.7 in.
Height (Metric)375 mm
Tolerance Metric±4 mL
TypeGraduated Cylinder
MaterialGlass
Capacity (Metric)500 mL
For Use With (Application)Used accuracy assured at the lowest graduation
IncludesHeavy Round Base
ScaleWhite ceramic graduations
Graduations5 mL
AutoclavableAutoclavable
Limit of Error+/- 4.0mL
Subdivision5mL
Unit SizeEA