Search Thermo Fisher Scientific
| Catalog Number | Sterility | Capacity (Metric) | Closure Type |
|---|---|---|---|
| FIS02-681-332 | Sterile | 2 mL | Attached Cap |
| FIS02-681-311 | Non-sterile | 0.6 mL | Snap Cap |
| FIS02-681-320 | Non-sterile | 1.5 mL | Snap Cap |
| FIS02-681-321 | Non-sterile | 2 mL | Snap Cap |
| FKS11906965 | Non-sterile | 2 mL | Snap Cap |
| FIS02-681-331 | Sterile | 1.5 mL | Attached Cap |