Search Fisher Scientific
| Catalog Number | Includes |
|---|---|
| 411-0125 also known as N411-0125NS | w/o tablet |
| FIS05-719-362 | w/o tablet |
Conduct analysis of drinking water with the Thermo Scientific™ Security-Snap™ Sterile Coliform Water Sample Bottle. It meets all EPA requirements for microbiology sample containers as specified by the EPA Manual for the Certification of Laboratories Analyzing Drinking Water. Made from non-toxic, non-autofluorescent polypropylene with a molded-in 100 ±2.5mL fill line. Vial features a Security-Snap flip-top lid, which provides tamper-evidence before and after sample collection, as well as protection against any possibility of accidental opening during transport.
Certified containers come with the following features: