LN2 Transfer Hose
LN<sub>2</sub> Transfer Hose
Actual product may vary
LN2 Transfer Hose
Provide the convenience, reassurance, and reliability required in the event of a power or mechanical failure with this LN2 transfer hose.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English) Exterior
40004016 ft.
40004004 ft.
Catalog number 4000401
Price (SGD)
641.00
EA
Add to cart
Length (English) Exterior:
6 ft.
Price (SGD)
641.00
EA
Add to cart
Provide the convenience, reassurance and reliability required in the event of a power or mechanical failure with Thermo Scientific™ Forma™ LN2 Transfer Hose. A must for critical storage applications, this system is for use with Forma ultra-low temperature freezers.
Features:
  • System is adjustable in 1°C increments to -150°C.
  • Digital display constantly indicates actual cabinet temperature.
  • Indicator lights when system is activated.
  • Liquid flow is shut off automatically when freezer door or lid is opened.
  • System is powered by rechargeable battery; low-battery light indicates when battery requires charging.
  • Additional indicator light illuminates when cylinder is empty.
Ordering Information:
  • Some models require freestanding system; others accept built-in systems.
  • Specify freezer model and voltage when ordering.
Includes:
All necessary hardware except LN2 cylinder (must be ordered separately)
Specifications
TypeLN2 Transfer Hose
Temperature Range-150°C to +1°C
For Use With (Equipment)LN2 Supply Tank
DefrostAutomatic
Length (English) Exterior6 ft.
Unit SizeEA