Cryo Vial Closure Color Coders
Actual product may vary
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Nunc
Cryo Vial Closure Color Coders
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
Catalog NumberColorProduct Line
NNC375884PinkNunc
Catalog number NNC375884
Price (SGD)
-
Color:
Pink
Product Line:
Nunc
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
Specifications
DescriptionNunc™ CryoColor Vial Closure Coder, PS
ColorPink
MaterialPolycarbonate
Product LineNunc
Quantity500/Pk., 2000/Cs.
Product TypeVial Closure
Unit SizePack