▸
Organic compounds
▸
Unclassified Organic Compounds
▸
Tetracyline hydrochloride, MP Biomedicals™
Element Search
Structure Search
MP Biomedicals
Tetracyline hydrochloride, MP Biomedicals™
Have Questions?
Change view
Quantity:
5 g
25 g
Catalog number
MPB0210301105
Price (SGD)
73.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (SGD)
73.00
Each
Add to cart
Specifications
Chemical Name or Material
Tetracyline hydrochloride
CAS
64-75-5
Color
Yellow
Molecular Formula
C22H25ClN2O8
Quantity
5 g
View more