Individual Stainless Steel Cylindrical Calibration Weights, Class M1
Product Image
Fisherbrand
Individual Stainless Steel Cylindrical Calibration Weights, Class M1
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWeight (Metric)
FKS1556515950 g
FKS15505169100 g
Catalog number FKS15565159
Price (SGD)
83.00
EA
Add to cart
Weight (Metric):
50 g
Price (SGD)
83.00
EA
Add to cart
  • Class M1 stainless steel calibration weight
  • In compliance with OIML R111
Specifications
ClassificationM1
MaterialStainless Steel
Weight (Metric)50 g
Mass50 g
ShapeCylindrical
Unit SizeEA