Check Order Status
Sample Pan for MB Series Moisture Analyzers
Actual product may vary
Fisherbrand
Sample Pan for MB Series Moisture Analyzers
Sample Pans for OHAUS™ MB series moisture analyzers.
Have Questions?
Change view
Catalog Number
Type
Quantity
FKS11865390
Reusable Sample Pan
3/Pk.
FKS11804242
Disposable Sample Pan
80/Pk.
2 Options
Catalog number
FKS11865390
Price (SGD)
521.00
PK
Add to cart
Type:
Reusable Sample Pan
Quantity:
3/Pk.
Price (SGD)
521.00
PK
Add to cart
Shape: round
Choose disposable or reusable
Specifications
For Use With (Equipment)
US: MB35,MB45,MB27,MB90,MB120 Moisture Analyzers
EU: MB27, MB25, MB90-GB, MB120-GB, MB23, MB120-EU
Quantity
3/Pk.
Type
Reusable Sample Pan