Shandon™ Kwik-Diff™ Stains
Shandon™ Kwik-Diff™ Stains
Actual product may vary
Fisherbrand
Shandon™ Kwik-Diff™ Stains
Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.
Have Questions?
Catalog NumberQuantityTypeIncludes
FKS104353103 x 500 mLKwik-Diff Stains500 mL Each: Fixative, Eosin, Methylene Blue
Catalog number FKS10435310
Price (SGD)
729.00
PK
Add to cart
Quantity:
3 x 500 mL
Type:
Kwik-Diff Stains
Includes:
500 mL Each: Fixative, Eosin, Methylene Blue
Price (SGD)
729.00
PK
Add to cart
  • Available in kit form or as individual components
  • Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori
Complete Kit (9990700) Includes:
Reagent 1: Fixative; Reagent 2: Eosin; Reagent 3: Methylene Blue
Specifications
Quantity3 x 500 mL
TypeKwik-Diff Stains
Includes500 mL Each: Fixative, Eosin, Methylene Blue
Unit SizePK