NanoCool™ Refrigerated Specimen Shipper
Actual product may vary
Therapak
NanoCool™ Refrigerated Specimen Shipper
No gel refrigerants required
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesQuantity
FIS22-130-418Sturdy transport boxUp to 36 standard 13 x 100mm tubes
FIS22-130-417Transport boxUp to 18 standard 13 x 100mm tubes
Catalog number FIS22-130-418
Price (SGD)
1,111.00
Case
Includes:
Sturdy transport box
Quantity:
Up to 36 standard 13 x 100mm tubes
Price (SGD)
1,111.00
Case
  • Portable unit maintains temperature from 2-8°C — ideal for overnight or international shipments
  • Weighs less and requires less space than conventional polystyrene boxes and ice packs, reducing shipping costs
  • Vacuum Insulated Panel (VIP) compartment transfers heat by evaporating small quantities of water at low pressure; a heat absorption method that's seven times more efficient than ice
  • Separate NanoCool cooler fits atop the VIP compartment and can be activated at room temperature with the push of a button
  • Small shipper holds up to 18 standard (13 × 100mm) transport tubes; large shipper holds up to 36 tubes
Specifications
QuantityUp to 36 standard 13 x 100mm tubes
IncludesSturdy transport box
TypePortable Unit
DurationFor overnight or international shipping
Unit SizeCase