IATA Shipping Boxes for Ambient Transport
Actual product may vary
Therapak
IATA Shipping Boxes for Ambient Transport
Ideal kit packing boxes for reference labs, clinical trials and pathology kits
Have Questions?
Catalog NumberDimensions (L x W x H) ExteriorType
FIS22-130-0506 x 4.25 x 4 in.Small Ambient IATA Transport Box
Catalog number FIS22-130-050
Price (SGD)
308.00
Case
Dimensions (L x W x H) Exterior:
6 x 4.25 x 4 in.
Type:
Small Ambient IATA Transport Box
Price (SGD)
308.00
Case
  • Each box has a specific tab-lock mechanism to keep boxes closed during transport
  • Ambient shipping boxes ideal for kit packaging and outbound shipping of diagnostic kits
  • Known as two-way shipper boxes, all ambient shipper boxes can be used for return transport to the laboratory in addition to outbound shipping
Specifications
Certifications/ComplianceMeets IATA Packing Instruction 650 dimensional requirements.
ApplicationsFor shipment of Biological Substance Category B specimens
SizeSmall
TypeSmall Ambient IATA Transport Box
Dimensions (L x W x H) Exterior6 x 4.25 x 4 in.
Dimensions (D x W x H) Interior5.25 x 4 x 3.75 in.
Unit SizeCase