95kPa Compliant Transport Bags
Actual product may vary
Therapak
95kPa Compliant Transport Bags
For use as a secondary water-tight receptacle
Have Questions?
Catalog NumberProduct TypeIncludes
FIS22-130-0232-pocket Transport BagAqui-Pak Pouch
Catalog number FIS22-130-023
Price (SGD)
752.00
Case
Product Type:
2-pocket Transport Bag
Includes:
Aqui-Pak Pouch
Price (SGD)
752.00
Case
For use as a secondary water-tight receptacle
  • Leak-resistant seal
  • Back pocket holds quarter-folded requisition forms
  • To transport multiple primary tubes, use the Aqui-Pak™ absorbent and tube separator
  • Easy-to-use — simply insert specimen tube and absorbent material into the front pocket, peel away the white tape liner, and fold down adhesive to cover the opening
  • For transport of multiple primary tubes, use the Aqui-Pak absorbent and tube separator with a 95 kPa bag
  • For transport of cryogenic cryoboxes in a frozen shipper, use the 12 × 15 in. (30.5 × 38.1cm), one-pocket bag, which accommodates up to 3 in. (7.6cm) tall cryoboxes
Specifications
DescriptionTwo-pocket bag with Aqui-Pak pouch
ColorClear with White Tape Liner
IncludesAqui-Pak Pouch
MaterialPlastic
Dimensions (L x W)11 x 7 in. (27.9 x 17.8cm)
Length (English)11 in.
Width (English)7 in.
Product Type2-pocket Transport Bag
Length (Metric)27.9 cm
Width (Metric)17.8 cm
Unit SizeCase