VACUETTE™ QUICKSHIELD Safety Tube Holder
VACUETTE™ QUICKSHIELD Safety Tube Holder
Actual product may vary
VACUETTE™ QUICKSHIELD Safety Tube Holder
Integrated safety mechanism is activated by an audible click
Have Questions?
Catalog NumberSize
FIS22-040-213Short standard
Catalog number FIS22-040-213
Price (SGD)
0.00
PK
Add to cart
Size:
Short standard
Price (SGD)
0.00
PK
Add to cart
Specifications
Certifications/ComplianceCE, OSHA
DisposableSingle-use
For Use With (Equipment)Vacuette™ Collection Needles
SizeShort standard
Storage Requirements4°C to 25°C
MaterialPlastic
Product TypeSafety Tube Holder
Unit SizePK