Temp-Gard™ Extreme Temperature Apron
Actual product may vary
PIP
Temp-Gard™ Extreme Temperature Apron
Black silicone apron protects torso and upper legs in extreme temperature conditions
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)
FIS19-149-65354 x 24 in. (137.2 x 61 cm)
FIS19-149-65142 x 24 in. (106.7 x 61 cm)
FIS19-149-65036 x 24 in. (91.4 x 61 cm)
FIS19-149-65248 x 24 in. (122 x 61 cm)
Catalog number FIS19-149-653
Price (SGD)
-
Dimensions (L x W):
54 x 24 in. (137.2 x 61 cm)
  • Protects from splashes
  • Multi-layered liquid-proof black silicone fabric
  • Impervious to cryogenic liquids and most other liquids and chemicals
  • Does not become slippery, even at extreme temperatures
  • Superior flexibility; will not stiffen
  • Easy to clean; will not absorb chemicals and will not stain
  • Quick-release buckles at neck and waist for added safety
  • Temperature range: -196° to +260°C (-321° to +500°F)
Cryogenic cleaning, cryogenic systems maintenance, conservation of food produce.
Specifications
TypeApron
SizeX-Large
MaterialSilicone
ColorBlack
Closure TypeTies
SterilityNon-sterile
Dimensions (L x W)54 x 24 in. (137.2 x 61 cm)
Length (English)54 in.
Length (Metric)137.2 cm
Width (English)24 in.
Width (Metric)61 cm
Unit SizeEach