StrateLine Autoclave Tape
Actual product may vary
Propper
StrateLine Autoclave Tape
Indicates status of sterlization jobs
Have Questions?
Catalog NumberTemperatureRangeMetric
FIS19-130-0341
Catalog number FIS19-130-0341
Price (SGD)
-
White tape with stripes that turn dark during steam sterilization to distinguish processed and unprocessed items.
  • Universal adhesive provides secure bonding and easy removal without traces of residue
  • 3 in. core
Steam Sterilization
Specifications
AutoclavableAutoclavable
Adhesive TypeRubber, Self-adhesive
CoatingSynthetic Resin
ColorWhite
FormatTape
MaterialCrepe Paper
Product TypeIndicator Tape
For Use With (Application)Steam Sterilization
Quantity48/Cs.
Length (English)180 ft.
Length (Metric)54.9 m
Width (English)0.75 in.
Diameter (English)3 in.
Diameter (Metric)7.62 cm
Unit SizeCase