ProCoat™ Premium PVC Coated Black Gloves with Interlock Liner - 18 in.
Actual product may vary
PIP
ProCoat™ Premium PVC Coated Black Gloves with Interlock Liner - 18 in.
Single dip construction with a straight cuff for additional length to protect the forearm from liquid run-off. Interlock liner dries quickly, sheds minimal lint, and enhances user comfort.
Have Questions?
Catalog number FIS19-120-1472
Price (SGD)
-
  • Single dip PVC gloves provide liquid and cut protection
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Black color helps to hide dirt and stains
Specifications
Coating MaterialPVC
ColorBlack
Exterior FinishNo Texture
HandSpecific
Product TypeDip-coated Gloves
SizeUniversal
For Use With (Application)Construction, Mining, Refining, and Petrochemicals
Glove Cuff StyleStraight
Liner MaterialInterlock Cotton
Series58-8060
Length (English)18 in.
Length (Metric)45.7 cm
Unit SizeDozen