50 Person Class A First Aid/Burn Kit
50 Person Class A First Aid/Burn Kit
Actual product may vary
50 Person Class A First Aid/Burn Kit
Class A first aid kits are intended to provide a basic range of products to deal with most common types of injuries encountered in the workplace including major wounds, minor wounds(cuts and abrasions), minor burns and eye injuries.
Have Questions?
Catalog NumberSufficient For
FIS19-090-86050 Persons
Catalog number FIS19-090-860
Price (SGD)
580.00
EA
Add to cart
Sufficient For:
50 Persons
Price (SGD)
580.00
EA
Add to cart
  • 50 person metal kit provides first aid for up to 50 people
  • Ideal for any small Lab setting to treat small cuts, burns, abrasions and injuries while at work
  • Can be wall mounted for ease of use
Kit Includes:
  • 1 x 3 in. Latex strips
  • 1/2 x 2.5 yd adhesive tape
  • Triple Antibiotic Ointement
  • Antiseptic wipes
  • CPR mask
  • BurnAid 4 x 4 dressing
  • Burn Cream with Lidocaine
  • Burn spray pump
  • Ice Pack
  • Eye pads
  • Eyewash 1 oz
  • Eyewash 8 oz
  • First aid handbook
  • Hand sanitizer
  • Dsposable vinyl gloves
  • 2 in. non-sterile conforming gauze
  • 4.5 in. wire scissors
  • 3 x 3 Sterile gauze pad
  • Abdominal sterile 5 x 9 in. pad
  • Triangular bandage
  • Liquid Skin 0.017 oz.
  • Calgonate 25gm
  • 1 x 3 BMD Plastic Strip
Specifications
For Use With (Application)Most common workplace injuries
Product TypeClass A First Aid/Burn Kit
Certifications/ComplianceMeets the 2015 ANSI Standards
Sufficient For50 Persons
Unit SizeEA