CleanPath™ Adhesive Mats
Actual product may vary
Berkshire
CleanPath™ Adhesive Mats
Berkshire CleanPath™ Adhesive Mats are the number one choice in cleanroom mats. Place CleanPath™ Adhesive Mats in cleanroom entry areas and change room entry areas to significantly decrease the transfer of foot-borne and wheel-borne contaminants including viable and non-viable particles.
Have Questions?
Catalog NumberDimensions (L x W)
FIS19-066-72136 x 18 in.
Catalog number FIS19-066-721
Price (SGD)
287.00
Case
Dimensions (L x W):
36 x 18 in.
Price (SGD)
287.00
Case
  • Excellent adhesive strength and cleanliness
  • Blue color allows differentiation for guarding against migration into critical areas
  • Adhesive coated surface film peels off easily for disposal
Specifications
Dimensions (L x W)36 x 18 in.
Quantity30 Sheets/Mat, 4 Mats/Cs.
ColorBlue
Length (English)36 in.
Length (Metric)91 cm
MaterialLDPE
TypeAdhesive Floor Mat
Width (English)18 in.
Width (Metric)46 cm
Unit SizeCase