▸
Personal Protective Equipment
▸
Safety Clothing
▸
Shoe and Boot Covers
▸
Controlled Environments Shoe and Boot Covers
▸
Polypropylene Shoe Cover - Non-skid
Actual product may vary
Keystone
Polypropylene Shoe Cover - Non-skid
Have Questions?
Catalog Number
Size
Weight (English)
Weight (Metric)
FIS19-055-175
Large
6 lb.
2.72 kg
Catalog number
FIS19-055-175
Price (SGD)
-
Size:
Large
Weight (English):
6 lb.
Weight (Metric):
2.72 kg
Specifications
Size
Large
Material
Polypropylene
Coating
Vinyl
Color
Blue
Sole Type
Non-skid
Packaging Quantity
150 Pairs
Weight (English)
6 lb.
Weight (Metric)
2.72 kg
Resistance
Fluid
Product Type
Controlled Environments Shoe Covers