Pleated Polypropylene Bouffant Cap
Actual product may vary
Keystone
Pleated Polypropylene Bouffant Cap
Comfortable, pleated bouffant caps feature an encapsulated elastic headband to keep hair in place. Available in a variety of sizes and colors.
Have Questions?
Catalog NumberSizeColor
FIS19-055-10324 in.White
Catalog number FIS19-055-103
Price (SGD)
26.00
Pack
Size:
24 in.
Color:
White
Price (SGD)
26.00
Pack
  • Polypropylene
  • 100% Latex Free
  • Economical Protection
  • Sizes Available: 21, 24, 28 in
  • Flexible, pleated design that stretches over the worker's head
  • Disposable design for fresh, hygienic use every time
Specifications
Product TypeBouffant
Size24 in.
MaterialPolypropylene
ColorWhite
DisposableSingle-use
Latex FreeYes
Model111NWI-10
SterilityNon-sterile
Quantity1000/Cs.
Diameter (English)24 in.
Diameter (Metric)61 cm
For Use With (Application)Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
StylePleated
Unit SizePack