Jomac™ Extra Heavyweight Terrycloth Heat Resistant Mitts
Actual product may vary
Wells Lamont
Jomac™ Extra Heavyweight Terrycloth Heat Resistant Mitts
Jomac™ Extra Heavyweight Terrycloth Heat Resistant Mitts feature a 9 in. gauntlet cuff for extended protection from high temperatures up to 450°F.
Have Questions?
Catalog NumberSize
FIS19-048-683Large
Catalog number FIS19-048-683
Price (SGD)
-
Size:
Large
  • Protection from temperatures up to 450°F
  • Constructed from extra heavy weight terrycloth
  • Cotton-lined hand
  • 9 in. gauntlet cuff
  • Loop-out
Specifications
Coating MaterialUncoated
ColorNatural
HandAmbidextrous
MaterialTerrycloth
Product TypeHeat-resistant Mitts
SizeLarge
Temperature (English) Max.450°F
Temperature (Metric) Max230°C
For Use With (Application)Bakeries, and Restaurant Establishments
Glove Cuff StyleGauntlet
Liner MaterialCotton
SeriesM937HR
Length (English)17 in.
Length (English) Cuff9 in.
Length (Metric)43.2 cm
Length (Metric) Cuff23 cm
Unit SizeDozen