▸
Personal Protective Equipment
▸
Respiratory Protection
▸
Face Masks
▸
BCR™ Cleanroom Masks
Actual product may vary
Berkshire
BCR™ Cleanroom Masks
Designed to protect cleanrooms and controlled environments from human contamination.
Have Questions?
Catalog Number
Includes
WidthEnglish
WidthMetric
FIS19-035-071
Catalog number
FIS19-035-071
Price (SGD)
-
Does not contain surfactants, fiberglass, glue, binders or thread
Provide filtration efficiency and long wearing comfort for operators
Soft, comfortable inside layer provides efficient particle and bacteria filtration and breathability
Hems, seams, and attachments are ultrasonically bound for strength and cleanliness
Cat. No 01-378 is individually wrapped and includes a clip to allow secure fastening behind head
Cat. No 19-035-071 comes bulk packaged and is extra wide reducing the amount of exposed skin
Specifications
Product Type
Cleanroom Face Mask
Color
White
Fastening Type
Elastic Straps
Material
Polypropylene
Style
Pleated
Cleanroom Class
Class 10 (ISO 4)
Construction
3-ply
Model
BM 1BEWO 12
Series
BCR Series